missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-Nornicotine, Dr. Ehrenstorfer

Product Code. 30319536
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319536 Supplier Dr. Ehrenstorfer Supplier No. DREC15651700
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Nornicotine
CAS 5746-86-1
Analyte or Component Names DL-Nornicotine
Shipping Condition +20°C
Molecular Formula C9 H12 N2
Certifications/Compliance ISO/IEC 17025
Synonym Nornicotine, Pyridine, 3-(2-pyrrolidinyl)-, (RS)-Nornicotine, (R,S)-Nornicotine, 2-(3-Pyridinyl)pyrrolidine, 3-(Pyrrolidin-2-yl)pyridine, rac-Nornicotine, Nicotine Imp. F (EP), Nicotine USP Related Compound F, Nicotine USP RC F, Nicotine, 1'-demethyl-, (+/-)- (8CI), Pyridine, 3-(2-pyrrolidinyl)-, (+/-)-, (+/-)-1'-Demethylnicotine, (+/-)-Nornicotine, Nicotine Related Compound F, Nicotine Impurity F as Racemate
InChI Formula InChI=1S/C9H12N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1,3,5,7,9,11H,2,4,6H2
SMILES C1CNC(C1)c2cccnc2
IUPAC Name 3-pyrrolidin-2-ylpyridine
Molecular Weight (g/mol) 148.2
Quantity 100 mg
Formula Weight 148.1 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.