missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-Nornicotine-d4 (100 ug/mL in Methanol), TRC

Product Code. 30395761
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395761 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30395761 Supplier Toronto Research Chemicals Supplier No. TRCKIT05625X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 66148-18-3
Molecular Formula C9 2H4 H8 N2
Quantity 5 x 1 mL
Synonym rac-Nornicotine-D4 (pyridine-D4), (±)-Nornicotine-D4, Pyridine-2,3,4,6-d4, 5-(2-pyrrolidinyl)-, Pyridine-2,3,4,6-d4, 5-(2-pyrrolidinyl)-, (±)-, 5-(2-Pyrrolidinyl)pyridine-2,3,4,6-d4
InChI Formula InChI=1S/C9H12N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1,3,5,7,9,11H,2,4,6H2/i1D,3D,5D,7D
SMILES [2H]c1nc([2H])c(C2CCCN2)c([2H])c1[2H]
IUPAC Name 2,3,4,6-tetradeuterio-5-pyrrolidin-2-ylpyridine
Molecular Weight (g/mol) 152.23
Formula Weight 152.1252
Concentration or Composition (by Analyte or Components) (±)-Nornicotine-D4 100 μg/mL
Chemical Name or Material (±)-Nornicotine-D4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.