missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-Nornicotine (1 mg/mL in Methanol), TRC

Product Code. 30391406
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30391406 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391406 Supplier Toronto Research Chemicals Supplier No. TRCKIT05601X1ML
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Nornicotine
CAS 5746-86-1
Molecular Formula C9 H12 N2
Quantity 1x1 mL
Synonym Nornicotine, Pyridine, 3-(2-pyrrolidinyl)-, Nicotine, 1'-demethyl-, (±)- (8CI), Pyridine, 3-(2-pyrrolidinyl)-, (±)-, (RS)-Nornicotine, (±)-1'-Demethylnicotine, (±)-Nornicotine, 2-(3-Pyridinyl)pyrrolidine, 3-(Pyrrolidin-2-yl)pyridine, rac-Nornicotine
InChI Formula InChI=1S/C9H12N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1,3,5,7,9,11H,2,4,6H2
SMILES C1CNC(C1)c2cccnc2
IUPAC Name 3-pyrrolidin-2-ylpyridine
Molecular Weight (g/mol) 148.2
Formula Weight 148.1
Concentration or Composition (by Analyte or Components) 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.