missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-N2-Nitroso-Anabasine N’-Beta-D-Glucuronide, TRC

Product Code. 30481460
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30481460 1 mg
30477036 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481460 Supplier Toronto Research Chemicals Supplier No. TRCN5242651MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1033078-12-4
Molecular Formula C16H22N3O7
Molecular Weight (g/mol) 367.35
Synonym 1-β-D-Glucopyranuronosyl-3-(1-nitroso-2-piperidinyl)pyridinium
SMILES C1CCN(C(C1)C2=C[N+](=CC=C2)[C@H]3C(C([C@@H](C(O3)C(=O)O)O)O)O)N=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H21N3O7/c20-11-12(21)14(16(23)24)26-15(13(11)22)18-6-3-4-9(8-18)10-5-1-2-7-19(10)17-25/h3-4,6,8,10-15,20-22H,1-2,5,7H2/p+1/t10?,11?,12-,13?,14?,15+/m0/s1
Chemical Name or Material (R,S)-N2-Nitroso-Anabasine N'-beta-D-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.