missing translation for 'onlineSavingsMsg'
Learn More

(R/S)-N-Deacetyl Colchiceine N-Trifluroracetate, TRC

Product Code. 30368718
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368718 Supplier Toronto Research Chemicals Supplier No. TRCD19890510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R/S)-N-Deacetyl Colchiceine N-Trifluroracetate
CAS 102491-70-3
Molecular Formula C21H20F3NO6
Quantity 10 mg
Synonym 2,2,2-Trifluoro-N-[5,6,7,9-tetrahydro-10-hydroxy-1,2,3-trimethoxy-9-oxobenzo[a]heptalen-7-yl]-acetamide
InChI Formula InChI=1S/C21H20F3NO6/c1-29-16-8-10-4-6-13(25-20(28)21(22,23)24)12-9-15(27)14(26)7-5-11(12)17(10)19(31-3)18(16)30-2/h5,7-9,13H,4,6H2,1-3H3,(H,25,28)(H,26,27)
SMILES COc1cc2CCC(NC(=O)C(F)(F)F)C3=CC(=O)C(=CC=C3c2c(OC)c1OC)O
IUPAC Name 2,2,2-trifluoro-N-(10-hydroxy-1,2,3-trimethoxy-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl)acetamide
Molecular Weight (g/mol) 439.38
Formula Weight 439.1243
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.