missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R,S)-Hydrobupropion Glucuronide, TRC
Specifications
Specifications
| Chemical Name or Material | (R,S)-Hydrobupropion Glucuronide |
| Analyte or Component Names | (R,S)-Hydrobupropion Glucuronide |
| Shipping Condition | Dry Pack |
| Molecular Formula | C19 H28 Cl N O7 |
| InChI Formula | InChI=1S/C19H28ClNO7/c1-9(21-19(2,3)4)15(10-6-5-7-11(20)8-10)27-18-14(24)12(22)13(23)16(28-18)17(25)26/h5-9,12-16,18,21-24H,1-4H3,(H,25,26)/t9-,12-,13-,14+,15-,16-,18+/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@H](NC(C)(C)C)[C@H](O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)O)c2cccc(Cl)c2 |
| IUPAC Name | (2S,3S,4S,5R,6R)-6-[(1R,2S)-2-(tert-butylamino)-1-(3-chlorophenyl)propoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 417.881 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?