missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-Hydrobupropion Glucuronide, TRC

Product Code. 30395601
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395601 2.5 mg 2.5mg
30398287 0.5 mg 0.5mg
30408752 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395601 Supplier Toronto Research Chemicals Supplier No. TRCH7145882.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R,S)-Hydrobupropion Glucuronide
Analyte or Component Names (R,S)-Hydrobupropion Glucuronide
Shipping Condition Dry Pack
Molecular Formula C19 H28 Cl N O7
InChI Formula InChI=1S/C19H28ClNO7/c1-9(21-19(2,3)4)15(10-6-5-7-11(20)8-10)27-18-14(24)12(22)13(23)16(28-18)17(25)26/h5-9,12-16,18,21-24H,1-4H3,(H,25,26)/t9-,12-,13-,14+,15-,16-,18+/m0/s1
Solution Type Neat
SMILES C[C@H](NC(C)(C)C)[C@H](O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)O)c2cccc(Cl)c2
IUPAC Name (2S,3S,4S,5R,6R)-6-[(1R,2S)-2-(tert-butylamino)-1-(3-chlorophenyl)propoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 417.881
Quantity 2.5 mg
Formula Weight 417.155
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.