missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-1-Tosyl Glycerol, TRC

Product Code. 30456925
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456925 1 g 1g
30453957 500 mg 500mg
30457234 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456925 Supplier Toronto Research Chemicals Supplier No. TRCT6640001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73073-07-1
Molecular Formula C10 H14 O5 S
Molecular Weight (g/mol) 246.28
IUPAC Name 2,3-dihydroxypropyl 4-methylbenzenesulfonate
SMILES Cc1ccc(cc1)S(=O)(=O)OCC(O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H14O5S/c1-8-2-4-10(5-3-8)16(13,14)15-7-9(12)6-11/h2-5,9,11-12H,6-7H2,1H3
Formula Weight 246.0562
Chemical Name or Material (R,S)-1-TosylGlycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.