missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-1-Methyl-3-nicotinoylpyrrolidine, TRC

Product Code. 17942165
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942165 100 mg 100mg
30134571 5 mg 5mg
17984055 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17942165 Supplier Toronto Research Chemicals Supplier No. TRCM323400100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125630-26-4
Molecular Formula C11 H14 N2 O
Molecular Weight (g/mol) 190.2417
Synonym Methanone, (1-methyl-3-pyrrolidinyl)-3-pyridinyl-, (RS)-1-Methyl-3-nicotinoylpyrrolidine ((RS)-MNP)
IUPAC Name (1-methylpyrrolidin-3-yl)-pyridin-3-ylmethanone
SMILES CN1CCC(C1)C(=O)c2cccnc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H14N2O/c1-13-6-4-10(8-13)11(14)9-3-2-5-12-7-9/h2-3,5,7,10H,4,6,8H2,1H3
Formula Weight 190.1106
Chemical Name or Material (RS)-1-Methyl-3-nicotinoylpyrrolidine ((RS)-MNP)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.