missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Lymphotoxin-alpha/TNF-beta Protein

Product Code. 18177730
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18177730

Brand: R&D Systems™ 211TBB010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cytotoxicity assay using L-929 mouse fibroblast cells in the presence of the metabolic inhibitor actinomycin D. Matthews, N. and M.L. Neale (1987) in Lymphokines and Interferons, A Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 221. The ED50 for this effect is 4-20 pg/mL.

Order Info

The product is shipped at ambient temperature. Upon receipt, store it immediately

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01374
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 4049
Molecular Weight (g/mol) M.W. (Observed): 17-19 kDa, reducing conditions; M.W. (theoretical): 19 kDa
Name Lymphotoxin-alpha/TNF-beta, Carrier Free
Preparation Method Reconstitute at 100μg/mL in PBS.
Quantity 10 μg
Source E. coli-derived human Lymphotoxin-alpha/TNF-beta protein Leu35-Leu205, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol LTA
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Form Powder
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Lymphotoxin-alpha/TNF-beta
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.