missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (R,R)-THC
GREENER_CHOICE

Product Code. 30001340
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30001340

Brand: Tocris Bioscience™ 1990/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent subtype-selective ligand; ERα agonist/ERβ antagonist

Non-steroidal, selective estrogen receptor ligand; agonist at ERα receptor (Ki = 9.0 nM) and antagonist at ERβ receptor (Ki = 3.6 nM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 138090-06-9
Quantity 10 mg
Molecular Formula C22H24O2
Formula Weight 320.42
Purity >98%
Molecular Weight (g/mol) 320.432
Chemical Name or Material (R, R)-THC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.