missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride, 97%

Product Code. 10017352 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10017352 1 g 1g
10279273 5 g 5g
10195980 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10017352 Supplier Thermo Scientific Acros Supplier No. 295810010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 138124-32-0 | C36H52ClMnN2O2 | 635.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138124-32-0
MDL Number MFCD02101664
InChI Key YRVXCOIDMFNGIJ-SEILFYAJSA-M
Synonym 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene manganese iii chloride r,r-jacobsen cat.
PubChem CID 131675872
IUPAC Name 2,4-ditert-butyl-6-[[[(1R,2R)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one;manganese;chloride
SMILES CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Cl-].[Mn]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 330°C to 332°C
Color Brown to Yellow
Elemental Analysis 96.0 % min.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C36H52ClMnN2O2
Quantity 1 g
Merck Index 15, 5300
Molecular Weight (g/mol) 635.21
Formula Weight 635.21
Physical Form Liquid
Chemical Name or Material (R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.