missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II), 98%

Product Code. 10127813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10127813 25 g Glass Bottle 25g
10576071 1 g Glass Bottle 1g
10676374 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10127813 Supplier Thermo Scientific Acros Supplier No. 329950250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 176763-62-5 | C36H52CoN2O2 | 603.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176763-62-5
Molecular Formula C36H52CoN2O2
Molecular Weight (g/mol) 603.76
MDL Number MFCD01631277
InChI Key XJWCTQVLJVAWMC-FAZMKCLSSA-N
Synonym r,r-n,n'-bis 3,5-di-tert-butylsalicylidene-1,2-cyclohexanediaminocobalt ii
PubChem CID 126963446
IUPAC Name cobalt;2,4-ditert-butyl-6-[[[(1R,2S)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one
SMILES CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Co]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >350°C
Color Brown to Orange
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 603.76
Physical Form Powder
Chemical Name or Material (R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II)
Show More Show Less
compliance-icons
Product Identifier
  • (1R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexnediaminocobalt(II)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.