missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | (R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide |
| CAS | 144222-34-4 |
| Molecular Formula | C21H22N2O2S |
| Quantity | 10 g |
| Synonym | (1R,2R)-(-)-N-(p-Tolylsulfonyl)-1,2-diphenylethylenediamine, (1R,2R)-(-)-N-p-Tosyl-1,2-diphenylethylenediamine, (1R,2R)-1-Amino-2-tosylamino-1,2-diphenylethane, (1R,2R)-N-p-Toluenesulfonyl-1,2-diphenylethylenediamine, (R,R)-1,2-Diphenyl-N-(p-toluenesulfonyl)ethylenediamine, (R,R)-N-(p-Toluenesulfonyl)-1,2-diphenylethylenediamine, N-[(1R,2R)-2-Amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide, TsDPEN, [(1R,2R)-1,2-Diphenyl-2-[[(4-tolyl)sulfonyl]amino]ethyl]amine |
| InChI Formula | InChI=1S/C21H22N2O2S/c1-16-12-14-19(15-13-16)26(24,25)23-21(18-10-6-3-7-11-18)20(22)17-8-4-2-5-9-17/h2-15,20-21,23H,22H2,1H3/t20-,21-/m1/s1 |
| SMILES | Cc1ccc(cc1)S(=O)(=O)N[C@@H]([C@H](N)c2ccccc2)c3ccccc3 |
| IUPAC Name | N-[(1R,2R)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide |
| Molecular Weight (g/mol) | 366.48 |
| Formula Weight | 366.1402 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction