missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine, 97%

Product Code. 11466470
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466470 500 mg 500mg
11456470 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466470 Supplier Thermo Scientific Alfa Aesar Supplier No. H27577.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90290-05-4 | C8H17NO2 | 159.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90290-05-4
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.229
MDL Number MFCD00210015
InChI Key HDXYTIYVVNJFLU-HTQZYQBOSA-N
Synonym 2r,5r-2,5-bis methoxymethyl pyrrolidine, r,r---2,5-bis methoxymethyl pyrrolidine, pyrrolidine, 2,5-bis methoxymethyl-, 2r,5r, 2r,5r-5-methoxymethyl pyrrolidin-2-yl methoxymethane, r,r---2,5-bis methoxymethyl-pyrrolidine, r,r-2,5-bis methoxymethyl pyrrolidine, 2beta,5alpha-bis methoxymethyl pyrrolidine
PubChem CID 2724974
IUPAC Name (2R,5R)-2,5-bis(methoxymethyl)pyrrolidine
SMILES COCC1CCC(N1)COC

Specifications

Boiling Point 106°C to 108°C (18 mmHg)
Refractive Index 1.449
Quantity 500 mg
Optical Rotation −8° (c=3 in Ethanol)
Formula Weight 159.23
Percent Purity 97%
Chemical Name or Material (R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.