missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-(+)-1,2-Diphenyl-1,2-ethanediol, 98+%, Thermo Scientific™

Product Code. 10791331 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
Glass bottle; 500mg
This item is not returnable. View return policy

Product Code. 10791331

Brand: Thermo Scientific Acros 279525000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 52340-78-0
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C14H14O2
Linear Formula C6H5CH(OH)CH(OH)C6H5
MDL Number MFCD00064254
Quantity 500mg
Beilstein 06, 970
Merck Index 14, 4777
Specific Rotation Condition +93° (20°C c=2.5,EtOH)
Specific Rotation + 93.00
Synonym r,r-+-hydrobenzoin, 1r,2r-1,2-diphenylethane-1,2-diol, r,r-hydrobenzoin, +-hydrobenzoin, unii-wv0i33nond, +-1r,2r-1,2-diphenylethane-1,2-diol, wv0i33nond, isohydrobenzoin, 1,2-ethanediol, 1,2-diphenyl-, 1r,2r-rel, +-dihydrobenzoin
InChI Key IHPDTPWNFBQHEB-ZIAGYGMSSA-N
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
IUPAC Name (1R,2R)-1,2-diphenylethane-1,2-diol
Molecular Weight (g/mol) 214.26
PubChem CID 853019
ChEBI CHEBI:50014
Formula Weight 214.26
Percent Purity 98+%
Chemical Name or Material (R,R)-(+)-1,2-Diphenyl-1,2-ethanediol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.