missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-1,2-Di(3-pyridyl)-1,2-ethanediamine tetrahydrochloride, 95%, ee 99%, Thermo Scientific™

Product Code. 15427535
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427535 250 mg 250mg
11405760 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427535 Supplier Thermo Scientific Alfa Aesar Supplier No. H25969.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1301706-42-2
Molecular Formula C12H18Cl4N4
Molecular Weight (g/mol) 360.104
MDL Number MFCD09265312
InChI Key GAHCWAYKHBFBHN-WVNQDYSFSA-N
Synonym r,r-1,2-di 3-pyridyl-1,2-ethanediamine tetrahydrochloride, r,r-1,2-bis 3-pyridyl-1,2-ethanediamine tetrahydrochloride, 1r,2r-1,2-bis pyridin-3-yl ethane-1,2-diamine tetrahydrochloride, r,r-1,2-di 3-pyridyl-1,2-ethanediamine tetrahydrochloride, ee
PubChem CID 45925680
IUPAC Name (1R,2R)-1,2-dipyridin-3-ylethane-1,2-diamine;tetrahydrochloride
SMILES C1=CC(=CN=C1)C(C(C2=CN=CC=C2)N)N.Cl.Cl.Cl.Cl

Specifications

Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 360.11
Percent Purity 95%
Assay ee 99%
Chemical Name or Material (R,R)-1,2-Di(3-pyridyl)-1,2-ethanediamine tetrahydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.