missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Quantikine Immunoassay Control Set 1005 for Human CCL7/MCP-3
Description
Quantikine Immunoassay Control Set 1005 for human CCL7/MCP-3 is intended for use as quantitative controls for the determination of cytokine concentrations in biological fluids.
- Applications: Control, ELISA
- Species Reactivity: Human
- Composition: Recombinant human cytokines at low, medium, and high concentrations, prepared in diluted porcine serum with preservatives, including ProClin 300
- Storage: Store lyophilized controls at 2°C to 8°C
- Stability: Stable for at least 6 Months from date of receipt when stored as directed
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Detectable Analytes | CCL7, MCP-3, MARC |
| Sample Type | Biological Fluids |
| Type | Immunoassay Control Set |
| Gene ID (Entrez) | 6354 |
| Intended Use | Control |
| Product Line | R&D Systems |
| Quantity | 1 Each |
| Source | Human |
| Form | Lyophilized |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?