missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 742 for Human HE4/WFDC2

Product Code. 18760539
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18760539 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18760539 Supplier R&D Systems™ Supplier No. QC102
  • ONLINE EXCLUSIVE PRICE
    £165.54
    £147.00 / Pack of 1
    Save £18.54
    11% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with catalog number DHE400

Quantikine Immunoassay Control Set 742 is designed for use with the human HE4/WFDC2 ELISA kit.

  • Species Reactivity: Human
  • Application: ELISA
  • Research Area: Cancer Biomarkers, Serine Proteases and Regulators
  • Storage: Store unopened product at 2°C to 8°C
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes HE4/WFDC2
Type Immunoassay Control
Gene ID (Entrez) 10406
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.