missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 726 for Human Enolase 2

Product Code. 18710948
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18710948 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18710948 Supplier R&D Systems™ Supplier No. QC83
  • ONLINE EXCLUSIVE PRICE
    £165.54
    £147.00 / Pack of 1
    Save £18.54
    11% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with catalog number DENL20

Quantikine™ Immunoassay Control Set 726 for Human Enolase 2 is designed for use as quantitative controls for the determination of cytokine concentrations in biological fluids.

  • Application: Control
  • Species Reactivity: Human
  • Form: Lyophilized
  • Content: Recombinant human cytokines at low, medium and high concentrations in a buffered protein solution with preservatives
  • Storage: Unopened product should be stored at 2°C to 8°C; stable for at least 6 months from date of receipt
  • Reconstitution: Reconstitute each vial with the volume of deionized or distilled water indicated on the product datasheet
  • Research Use Only: This product is for research use only
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Enolase 2 (Neuron-specific Enolase)
Type Immunoassay Control
Gene ID (Entrez) 2026
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.