missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ R-Phycoerythrin, Pyridyldisulfide Derivative
Description
The pyridyldisulfide derivative of R-phycoerythrin (R-PE) can be reacted directly with thiolated antibodies, enzymes and other biomolecules to form a disulfide linkage.
Specifications
Specifications
| Concentration | 2 mg/mL |
| Quantity | 1 mL |
| Product Type | Pyridyldisulfide Derivative of R-phycoerythrin |
| Content And Storage | Store in refrigerator (2–8°C) and protect from light. |
| Shipping Condition | Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?