missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ R-Phycoerythrin, Biotin-XX Conjugate

Product Code. 10413832
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10413832 0.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10413832 Supplier Invitrogen™ Supplier No. P811

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

R-Phycoerythrin, Biotin-XX Conjugate

R-phycoerythrin biotin-XX conjugate consists of the exceptionally bright phycobiliprotein, R-PE conjugated with biotin-XX, where the biotin is well separated from the point of attachment by a fourteen-atom spacer.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 4 mg/mL
Conjugate Biotin
Quantity 0.5 mL
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Shipping Condition Wet Ice
Form Liquid
Protein Family Other Proteins
Product Type Biotin Conjugate (R-PE)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.