missing translation for 'onlineSavingsMsg'
Learn More

(R)-Nornicotine, TRC

Product Code. 30418101
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Product Code. Quantity
30418101 1 g
30412264 500 mg
30414089 100 mg
3 options
This item is not returnable. View return policy

Product Code. 30418101

Brand: Toronto Research Chemicals TRCN7570051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7076-23-5
Formula Weight 148.1
InChI Formula InChI=1S/C9H12N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1,3,5,7,9,11H,2,4,6H2/t9-/m1/s1
IUPAC Name 3-[(2R)-pyrrolidin-2-yl]pyridine
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.2
SMILES C1CN[C@H](C1)c2cccnc2
Synonym 3-(2R)-2-pyrrolidinylpyridine, (+)-1'-demethylnicotine, (R)-3-(2-pyrrolidinyl)pyridine, (+)-1'-Demethylnicotine, (+)-2-(3'-Pyridyl)pyrrolidine, (+)-Nornicotine, (R)-(+)-Nornicotine, (R)-(+)-Nornicotine
Chemical Name or Material (R)-Nornicotine

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.