missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine, 97%, Thermo Scientific Chemicals

Product Code. 11359445
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359445 1 g 1g
11379445 250 mg 250mg
11369445 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359445 Supplier Thermo Scientific Alfa Aesar Supplier No. L18659.03
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 153837-28-6 | Molecular Formula: C18H30N2 | Molecular Weight: 274.452 g/mol

(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine is a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Store under inert gas. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153837-28-6
Molecular Formula C18H30N2
Molecular Weight (g/mol) 274.452
MDL Number MFCD06795639
InChI Key RUWFXOINQANLGF-KRWDZBQOSA-N
Synonym r-2,2-dimethyl-n-1-phenyl-2-piperidin-1-yl ethyl propan-1-amine, r---n-neopentyl-1-phenyl-2-1-piperidinyl ethylamine, 1r-1-phenyl-2-piperidylethyl 2,2-dimethylpropyl amine, 2,2-dimethylpropyl 1r-1-phenyl-2-piperidin-1-yl ethyl amine, 1r-n-2,2-dimethylpropyl-1-phenyl-2-piperidinoethanamine, 2,2-dimethyl-n-1r-1-phenyl-2-piperidin-1-ylethyl propan-1-amine
PubChem CID 7577799
IUPAC Name 2,2-dimethyl-N-[(1R)-1-phenyl-2-piperidin-1-ylethyl]propan-1-amine
SMILES CC(C)(C)CNC(CN1CCCCC1)C2=CC=CC=C2

Specifications

Melting Point 36°C to 40°C
Quantity 1 g
UN Number UN3259
Beilstein 6957993
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Optical Rotation −90° (c=1 in Methanol)
Formula Weight 274.45
Percent Purity 97%
Chemical Name or Material (R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.