missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%, Thermo Scientific Chemicals

Product Code. 11365505
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
Unit Size:
100mg
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365505 100 mg 100mg
11345505 500 mg 500mg
11355505 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11365505 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' L19685.MC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine acts as a ligand used in the enantioselective vinylation of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine acts as a ligand used in the enantioselective vinylation of aldehydes.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55700-44-2
Molecular Formula C26H28FeNP
Molecular Weight (g/mol) 441.34
MDL Number MFCD00075286
InChI Key RCAFPSZHKFOLEE-KVEBOLLKNA-N
IUPAC Name (R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine
SMILES [Fe].c1cccc1.C[C@@H](N(C)C)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 141°C to 143°C
Quantity 100 mg
Optical Rotation −355° (c=0.6 in Ethanol)
Formula Weight 441.34
Percent Purity 97%
Chemical Name or Material (R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.