missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine, 97%, Thermo Scientific Chemicals

Product Code. 11355495
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355495 250 mg 250mg
11345495 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11355495 Supplier Thermo Scientific Alfa Aesar Supplier No. L19679.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31886-58-5
Molecular Formula C14H19FeN
Molecular Weight (g/mol) 257.16
MDL Number MFCD00066273
IUPAC Name (R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine
SMILES [Fe].c1cccc1.C[C@@H](N(C)C)c1cccc1

Specifications

Density 1.222
Boiling Point 120°C to 121°C (0.7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.589
Quantity 250 mg
Optical Rotation +13° (c=1 in Ethanol, 5H)
Formula Weight 257.16
Percent Purity 97%
Chemical Name or Material (R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.