missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-(-)-N-Methyl-1-phenyl-2-(1-pyrrolidinyl)ethylamine, tech. 90%

Product Code. 11399435
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11399435

Brand: Alfa Aesar™ L18657.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 136329-39-0
Molecular Formula C13H20N2
Molecular Weight (g/mol) 204.317
MDL Number MFCD01318625
InChI Key ZINZYRWMDNKTBY-ZDUSSCGKSA-N
Synonym r-n-methyl-1-phenyl-2-pyrrolidin-1-yl ethanamine, r---n-methyl-1-phenyl-2-1-pyrrolidinyl ethylamine, methyl 1r-1-phenyl-2-pyrrolidin-1-yl ethyl amine, 1r-1-phenyl-2-pyrrolidinylethyl methylamine, 1-r-beta-methylamino phenethyl pyrrolidine, 1-pyrrolidineethanamine,n-methyl-a-phenyl-, ar, 1r-n-methyl-1-phenyl-2-1-pyrrolidinyl ethanamine, 1r-n-methyl-1-phenyl-2-pyrrolidin-1-yl-ethanamine, 1r-n-methyl-1-phenyl-2-pyrrolidin-1-ylethanamine
PubChem CID 7577779
IUPAC Name (1R)-N-methyl-1-phenyl-2-pyrrolidin-1-ylethanamine
SMILES CNC(CN1CCCC1)C2=CC=CC=C2

Specifications

Density 0.974
Refractive Index 1.5245
Quantity 1 g
UN Number UN2735
Beilstein 4842022
Sensitivity Air sensitive
Optical Rotation −78° (Neat)
Formula Weight 204.32
Percent Purity 90%
Grade Technical
Chemical Name or Material (R)-(-)-N-Methyl-1-phenyl-2-(1-pyrrolidinyl)ethylamine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.