missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-FMOC-α-Methylvaline, 98%, 98% ee, Thermo Scientific Chemicals

Product Code. 11985561 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11985561 100 mg 100mg
11955561 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985561 Supplier Thermo Scientific Acros Supplier No. 441141000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 616867-28-8 | Molecular Formula: C21H23NO4 | Molecular Weight: 353.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616867-28-8 , 1634-04-4
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD12031696
InChI Key AWEZXIRZNQCCNN-OAQYLSRUSA-N
Synonym r-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid, l-isovaline, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl, r-n-fmoc-alpha-methylvaline, fmoc-r-2-amino-2,3-dimethylbutanoicacid, fmoc-r-2-amino-2,3-dimethylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-l-isovaline, r-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid, r-2-9h-fluorene-9-ylmethoxycarbonylamino-2,3-dimethylbutyric acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid
PubChem CID 11210384
SMILES CC(C)C(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 53°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 mg
Specific Rotation -8.6
Residual Solvents 10% max. (MTBE)
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2,3-dimethylbutanoic acid
Formula Weight 353.43
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-N-FMOC-α-Methylvaline, 98% ee
Show More Show Less
compliance-icons
Product Identifier
  • (R)-N-FMOC-alpha-Methylvaline
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.