missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-FMOC-(2-Furyl)alanine, 95%, 98% ee, Thermo Scientific™

Product Code. 10282472 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10282472 1 g Glass bottle 1g
10743603 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10282472 Supplier Thermo Scientific Acros Supplier No. 362490010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 220497-85-8
Molecular Formula C22H19NO5
Molecular Weight (g/mol) 377.40
MDL Number MFCD01311747
InChI Key AJXDCHXGNUFBRC-UHFFFAOYSA-N
Synonym fmoc-d-2-furylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-d-ala-oh, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-3-2-furyl-d-alanine, n-fmoc-3-2-furyl-d-alanine, fmoc-beta-2-furyl-d-ala-oh hplc
PubChem CID 16211163
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid
SMILES OC(=O)C(CC1=CC=CO1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 121.6°C
Color White to Yellow
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 377.39
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (R)-N-FMOC-(2-Furyl)alanine, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.