missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-N-BOC-2-Piperidinecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10346444
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
28697-17-8
C11H19NO4
229.27
MFCD00237380
JQAOHGMPAAWWQO-MRVPVSSYSA-N
r-+-n-boc-2-piperidinecarboxylic acid, boc-d-hopro-oh, boc-d-pipecolic acid, r-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-d-pip-oh, d-n-boc-pipecolic acid, r-1-n-boc-pipecolinic acid, r-n-boc-piperidine-2-carboxylic acid, +-n-boc-r-pipecolinic acid, r-n-boc-2-piperidinecarboxylic acid
688618
(2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CCCCC1C(=O)O
This item is not returnable. View return policy
28697-17-8
C11H19NO4
229.27
MFCD00237380
JQAOHGMPAAWWQO-MRVPVSSYSA-N
r-+-n-boc-2-piperidinecarboxylic acid, boc-d-hopro-oh, boc-d-pipecolic acid, r-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-d-pip-oh, d-n-boc-pipecolic acid, r-1-n-boc-pipecolinic acid, r-n-boc-piperidine-2-carboxylic acid, +-n-boc-r-pipecolinic acid, r-n-boc-2-piperidinecarboxylic acid
688618
(2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CCCCC1C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 116°C to 119°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Specific Rotation +62.0° to +65.0° (20°C, 589nm) (c=1%, acetic acid)
Formula Weight 229.27
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-N-BOC-2-Piperidinecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-N-BOC-2-Piperidinecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'