missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-N-Allyl-1-phenylethylamine hydrochloride, 95%, Thermo Scientific™

Product Code. 10011821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10011821 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10011821 Supplier Thermo Scientific Acros Supplier No. 337850010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 204587-86-0
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C11H15N·HCl
Linear Formula C6H5CH(CH3)NHCH2CHCH2·HCl
MDL Number MFCD01862108
Quantity 1 g
Synonym r--n-allyl-1-phenylethylamine hydrochloride, 1r-1-phenylethyl prop-2-en-1-yl amine hydrochloride, r-+-n-2-allyl phenylethylamine, r-n-1-phenylethyl prop-2-en-1-amine hydrochloride
InChI Key BAFILFKLBRDSLS-HNCPQSOCSA-N
SMILES CC(C1=CC=CC=C1)NCC=C.Cl
IUPAC Name N-[(1R)-1-phenylethyl]prop-2-en-1-amine;hydrochloride
Molecular Weight (g/mol) 197.71
PubChem CID 53393158
Formula Weight 197.71
Percent Purity 95%
Physical Form Chunks and/or Powder
Chemical Name or Material (R)-(+)-N-Allyl-1-phenylethylamine hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.