missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-Acetyl-α-allylglycine, 98%, 98% ee, Thermo Scientific™

Catalog Number 11995521
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
121786-40-1
C7H11NO3
157.17
QTNLDKHXFVSKCF-ZCFIWIBFSA-N
2r-2-acetamidopent-4-enoic acid, ambotzaaa1953, r-n-acetyl-alpha-allylglycine ee, r, ?-2-acetylamino-4-pentenoic acid, rac-2r-2-acetamidopent-4-enoic acid, r-n-alpha-acetyl-allylglycineee
41097732
(2R)-2-acetamidopent-4-enoic acid
CC(=O)NC(CC=C)C(=O)O
This item is not returnable. View return policy
121786-40-1
C7H11NO3
157.17
QTNLDKHXFVSKCF-ZCFIWIBFSA-N
2r-2-acetamidopent-4-enoic acid, ambotzaaa1953, r-n-acetyl-alpha-allylglycine ee, r, ?-2-acetylamino-4-pentenoic acid, rac-2r-2-acetamidopent-4-enoic acid, r-n-alpha-acetyl-allylglycineee
41097732
(2R)-2-acetamidopent-4-enoic acid
CC(=O)NC(CC=C)C(=O)O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 101°C to 106°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 100 mg
Specific Rotation -8.3
Formula Weight 157.17
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-N-Acetyl-α-allylglycine, 98% ee
Show More Show Less
Product Identifier
  • (R)-N-Acetyl-alpha-allylglycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'