missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse FGF acidic/FGF1 DuoSet ELISA

Product Code. 30020944
Click to view available options
Quantity:
5 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30020944

Brand: R&D Systems™ DY468605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sufficient for at least five 96-well plates

The Mouse FGF acidic DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse FGF acidic/FGF1.
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes FGF acidic
Assay ELISA Development Kit
Conjugate Biotin
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well microplate (sold separately)
For Use With (Application) ELISA
Gene ID (Entrez) 14164
Host Species Mouse
Measuring Range 12.5-800pg/mL
Quantity 5 Plates
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym AFGF, alpha, alpha-ECGF, beta-ECGF, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGF acidic, FGF-1, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.