missing translation for 'onlineSavingsMsg'
Learn More

R-Mirtazapine, TRC

Product Code. 30471107
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. Quantity unitSize
30471107 25 mg 25mg
30481357 5 mg 5mg
30465909 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 30471107

Brand: Toronto Research Chemicals TRCM36499525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61364-37-2
Formula Weight 265.1579
InChI Formula InChI=1S/C17H19N3/c1-19-9-10-20-16(12-19)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)20/h2-8,16H,9-12H2,1H3/t16-/m0/s1
Molecular Formula C17 H19 N3
Molecular Weight (g/mol) 265.35
SMILES CN1CCN2[C@@H](C1)c3ccccc3Cc4cccnc24
Synonym Pyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, 1,2,3,4,10,14b-hexahydro-2-methyl-, (14bR)-, Pyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, 1,2,3,4,10,14b-hexahydro-2-methyl-, (R)-, (14bR)-1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, (-)-Mirtazapine, (R)-6-Azamianserin, (R)-Mirtazapine, (R)-Org 3770, Org 44-19, R-(-)-Mirtazapine, R-Mirtazapine
Chemical Name or Material (R)-Mirtazapine

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.