missing translation for 'onlineSavingsMsg'
Learn More

(R)-α-Methyltyrosine, 98%, 98% ee, Thermo Scientific™

Product Code. 11905441 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11905441 250 mg 250mg
11975431 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11905441 Supplier Thermo Scientific Acros Supplier No. 441242500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 672-86-6
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
InChI Key NHTGHBARYWONDQ-SNVBAGLBSA-N
Synonym r-alpha-methyltyrosine, alpha-methyl-d-tyr, r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid, 2r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid, alpha-methyl-d-tyrosine, metirosinum inn-latin, metirosina inn-spanish, l-tyrosine, alpha-methyl-,-, d +-metyrosine, metyrosine usan
PubChem CID 12650
IUPAC Name (2R)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid
SMILES CC(CC1=CC=C(C=C1)O)(C(=O)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >211°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Specific Rotation -18
Formula Weight 195.22
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-α-Methyltyrosine, 98% ee
Show More Show Less
Product Identifier
  • (R)-alpha-Methyltyrosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.