missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-alpha-Methoxyphenylacetic acid, 99%

Product Code. p-3778981 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10490301

Brand: Thermo Scientific Acros 224470010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3966-32-3 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3966-32-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004250
InChI Key DIWVBIXQCNRCFE-MRVPVSSYSA-N
Synonym r---alpha-methoxyphenylacetic acid, r-2-methoxy-2-phenylacetic acid, 2r-2-methoxy-2-phenylacetic acid, r-methoxyphenylacetic acid, r-methoxy phenyl acetic acid, methoxy phenyl acetic acid #, r---.alpha.-methoxyphenylacetic acid, benzeneacetic acid, .alpha.-methoxy-, r, o-methyl-d-mandelic acid, ksc496q9j
PubChem CID 2724294
IUPAC Name (2R)-2-methoxy-2-phenylacetic acid
SMILES COC(C1=CC=CC=C1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 64°C to 67°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5CH(OCH3)COOH
Quantity 1 g
Beilstein 10, 195
Specific Rotation Condition −160.40° (20°C c=1,methanol)
Specific Rotation -160.4
Formula Weight 166.18
Percent Purity 99%
Physical Form Fine Crystalline Powder or Needles
Chemical Name or Material (R)-(-)-α-Methoxyphenylacetic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.