missing translation for 'onlineSavingsMsg'
Learn More

R-(-)-Mecamylamine Hydrochloride, TRC

Product Code. 30482302
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482302 50 mg 50mg
30481621 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30482302 Supplier Toronto Research Chemicals Supplier No. TRCM20259050MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107596-31-6
Molecular Formula C11 H21 N . Cl H
Molecular Weight (g/mol) 203.75
Synonym Bicyclo[2.2.1]heptan-2-amine, N,2,3,3-tetramethyl-, hydrochloride, (1S,2R,4R)- (9CI), Bicyclo[2.2.1]heptan-2-amine, N,2,3,3-tetramethyl-, hydrochloride, (1S-exo)-, (-)-Mecamylamine hydrochloride, NIH 11009, R-(-)-Mecamylamine hydrochloride
IUPAC Name (1R,3R,4S)-N,2,2,3-tetramethylbicyclo[2.2.1]heptan-3-amine;hydrochloride
SMILES Cl.CN[C@]1(C)[C@H]2CC[C@H](C2)C1(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H21N.ClH/c1-10(2)8-5-6-9(7-8)11(10,3)12-4;/h8-9,12H,5-7H2,1-4H3;1H/t8-,9+,11-;/m1./s1
Formula Weight 203.1441
Chemical Name or Material R-(-)-Mecamylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.