missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Mandelic Acid, TRC

Product Code. 30133738
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133738 10 g 10g
30012543 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133738 Supplier Toronto Research Chemicals Supplier No. TRCM16253010G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-71-2
Molecular Formula C8 H8 O3
Molecular Weight (g/mol) 152.15
Synonym (2R)-Hydroxyphenylacetic Acid, Sertraline Hydrochloride Imp. E (EP), (R)-Mandelic acid, Benzeneacetic acid, α-hydroxy-, (αR)-, Benzeneacetic acid, α-hydroxy-, (R)-, Mandelic acid, D- (8CI), (αR)-α-Hydroxybenzeneacetic acid, (-)-(R)-Mandelic acid, (-)-D-Mandelic acid, (-)-Mandelic acid, (-)-α-Hydroxyphenylacetic acid, (2R)-2-Hydroxy-2-phenylacetic acid, (R)-(-)-Hydroxyphenylacetic acid, (R)-(-)-Mandelic acid, (R)-2-Hydroxy-2-phenylacetic acid, (R)-Amygdalic acid, (R)-Hydroxy(phenyl)acetic acid, (R)-α-Hydroxybenzeneacetic acid, (R)-α-Hydroxyphenylacetic acid, D(-)-Mandelic acid, D-2-Phenylglycolic acid, D-Mandelic acid
IUPAC Name (2R)-2-hydroxy-2-phenylacetic acid
SMILES O[C@@H](C(=O)O)c1ccccc1

Specifications

InChI Formula InChI=1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)/t7-/m1/s1
Formula Weight 152.0473
Percent Purity >95%
Quantity 10 g
Chemical Name or Material (R)-Mandelic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.