missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Mandelic acid, 98%

Product Code. 11417836
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417836 25 g 25g
11407836 5 g 5g
11427836 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417836 Supplier Thermo Scientific Alfa Aesar Supplier No. A14767.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-71-2 | C8H8O3 | 152.15 g/mol

(R)-(-)-Mandelic acid, is used as a antiseptic ingredient particularly against urinary tract infections.Mandelic acid and its derivatives are used to apply the dual activities as an antibacterial agent and as an antiaging agent. It is used as an intermediate for the synthesis of target molecules for other applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-Mandelic acid, is used as a antiseptic ingredient particularly against urinary tract infections.Mandelic acid and its derivatives are used to apply the dual activities as an antibacterial agent and as an antiaging agent. It is used as an intermediate for the synthesis of target molecules for other applications.

Solubility
It is partly soluble in water, freely soluble in isopropyl and ethyl alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Stable under ordinary conditions. Incompatabile to strong bases, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-71-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064251
InChI Key IWYDHOAUDWTVEP-SSDOTTSWSA-N
Synonym r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
PubChem CID 11914
ChEBI CHEBI:17656
IUPAC Name (2R)-2-hydroxy-2-phenylacetic acid
SMILES O[C@@H](C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 130°C to 134°C
Beilstein 2691094
Solubility Information It is partly soluble in water,freely soluble in isopropyl and ethyl alcohol.
Formula Weight 152.15
Percent Purity 98%
Quantity 25 g
Optical Rotation -153° (c=5 in Water)
Sensitivity Light sensitive
Chemical Name or Material (R)-(-)-Mandelic acid
compliance-icons
Product Identifier
  • D-(-)-Mandelic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: D-(-)-Mandelic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.