missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-Ketorolac, MedChemExpress
Description
(R)-Ketorolac is the R-enantiomer of Ketorolac, shows potent analgesic activity, reduces ulcerogenic potential. (R)-Ketorolac is inactive on COX.
Specifications
Specifications
| Chemical Name or Material | (R)-Ketorolac |
| CAS | 66635-93-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C15H13NO3 |
| Quantity | 5 mg |
| Synonym | (+)-Ketorolac |
| Solubility Information | DMSO : ≥ 100 mg/mL (391.74 mM) |
| SMILES | O=C(C1=CC=CC=C1)C2=CC=C3N2CC[C@H]3C(O)=O |
| Molecular Weight (g/mol) | 255.27 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?