missing translation for 'onlineSavingsMsg'
Learn More

(R)-Jorgensen-Hayashi Catalyst, TRC

Product Code. 30393494
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393494 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393494 Supplier Toronto Research Chemicals Supplier No. TRCJ2120201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-Jorgensen-Hayashi Catalyst
CAS 943757-71-9
Molecular Formula C20H27NOSi
Quantity 1 g
Synonym (2R)-2-[Diphenyl[(trimethylsilyl)oxy]methyl]pyrrolidine, (R)-Diphenylprolinol Trimethyl Silyl Ether, (R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol Trimethylsilyl Ether
InChI Formula InChI=1S/C20H27NOSi/c1-23(2,3)22-20(19-15-10-16-21-19,17-11-6-4-7-12-17)18-13-8-5-9-14-18/h4-9,11-14,19,21H,10,15-16H2,1-3H3/t19-/m1/s1
SMILES C[Si](C)(C)OC([C@H]1CCCN1)(c2ccccc2)c3ccccc3
IUPAC Name [diphenyl-[(2R)-pyrrolidin-2-yl]methoxy]-trimethylsilane
Molecular Weight (g/mol) 325.52
Formula Weight 325.1862
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.