missing translation for 'onlineSavingsMsg'
Learn More

R(+)-IAA-94, MedChemExpress

Product Code. 16481497
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481497 50 mg 50mg
16455298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481497 Supplier Medchem Express Supplier No. HY12693
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress R(+)-IAA-94 (R(+)-Methylindazone) is a potent indanyloxyacetic acid blocker of epithelial chloride channels. R(+)-IAA-94 inhibits Nef-sdAb19 (single-domain antibody) interaction and binds to negative factor (Nef).

R(+)-IAA-94 (R(+)-Methylindazone) is a potent indanyloxyacetic acid blocker of epithelial chloride channels. R(+)-IAA-94 inhibits Nef-sdAb19 (single-domain antibody) interaction and binds to negative factor (Nef).

Specifications

Chemical Name or Material R(+)-IAA-94
CAS 54197-31-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H18Cl2O4
Quantity 50 mg
Synonym R(+)-Methylindazone
Solubility Information DMSO : ≥ 100 mg/mL (279.93 mM) ∣H2O : 0.1 mg/mL (0.28 mM; Need ultrasonic)
SMILES O=C(O)COC1=CC2=C(C([C@@](C)(C3CCCC3)C2)=O)C(Cl)=C1Cl
Molecular Weight (g/mol) 357.23
Formula Weight 357.23
Percent Purity 98.07%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.