Learn More
R&D Systems™ Human SMVT/SLC5A6 (NP_066918) VersaClone cDNA
Product Code. 18764988
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18764988 10 μg 10µg
1 options
Product Code. 18764988 Supplier R&D Systems™ Supplier No. RDC1052
Sodium-Dependent Multivitamin Transporter
The Human SMVT/SLC5A6 (NP_066918) VersaClone cDNA clone contains the complete ORF for SMVT/SLC5A6, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SMVT/SLC5A6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans