missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Klotho beta DuoSet ELISA

Product Code. 30000552
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 30000552

Brand: R&D Systems™ DY588905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sufficient for at least five 96-well plates

The Human Klotho beta DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Klotho beta.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.20 to 2000 pg/mL
Detectable Analytes Klotho beta
Assay ELISA Development Kit
Conjugate Biotin
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well microplate (sold separately)
Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Gene ID (Entrez) 152831
Gene Symbol KLB
Host Species Human
Measuring Range 31.2-2,000pg/mL
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213
Target Klotho beta
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.