Learn More
R&D Systems™ Human IL-18 R beta/IL-1 R7 (NP_003844) VersaClone cDNA
Product Code. 18724619
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18724619 10 μg 10µg
1 options
Product Code. 18724619 Supplier R&D Systems™ Supplier No. RDC1118
  • ONLINE EXCLUSIVE PRICE
    £301.00
    £284.55 / 10µg
    Save £16.45
    5% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
Interleukin 18 Receptor beta
The Human IL-18 R beta/IL-1 R7 (NP_003844) VersaClone cDNA clone contains the complete ORF for IL-18 R beta/IL-1 R7/ACPL, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The IL-18 R beta/IL-1 R7/ACPL insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans