missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD31/PECAM-1 Quantikine ELISA Kit

Product Code. 18721804
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18721804

Brand: R&D Systems™ DCD310

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human CD31/PECAM-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CD31/PECAM-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CD31/PECAM-1 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), saliva (50 ul), urine (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.31 - 20ng/mL
Assay Sensitivity 0.078ng/mL
Detectable Analytes CD31/PECAM-1
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human CD31
For Use With (Application) ELISA
Gene ID (Entrez) 5175
Host Species Human
Measuring Range 0.313-20ng/mL
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Saliva (50uL),Urine (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.