missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL18/PARC Quantikine ELISA Kit

Product Code. 18761784
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18761784

Brand: R&D Systems™ DCL180B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human CCL18/PARC Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CCL18/PARC Quantikine ELISA Kit quantifies human CCL18/PARC in cell culture supernates (25 ul), tissue lysates (10 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (25 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 18.80 - 1200pg/mL
Assay Sensitivity 1.77pg/mL
Detectable Analytes CCL18/PARC
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Lysates, Urine
Specificity Natural and Recombinant Human CCL18
For Use With (Application) ELISA
Gene ID (Entrez) 6362
Host Species Human
Measuring Range 18.8-1,200pg/mL
Quantity 1 Kit
Sample Volume Cell Culture Supernates (25uL),Tissue Lysates (10uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Alternative macrophage activation-associated CC chemokine 1, AMAC-1, AMAC1CC chemokine ligand 18, AMAC-1 Small-inducible cytokine A18, chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated), CKb7, DC-CK1, DC-CK1CC chemokine PARC, DCCK1Macrophage inflammatory protein 4, Dendritic cell chemokine 1, MIP4, MIP-4, MIP-4 C-C motif chemokine 18, PARC, PARCPulmonary and activation-regulated chemokine, SCYA18chemokine (C-C), dendritic, small inducible cytokine A18, small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary andactivation-regulated
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.