missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Gossypol acetic acid, MedChemExpress

Product Code. 16426548
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16426548

Brand: Medchem Express HY15464A10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (R)-(-)-Gossypol acetic acid (AT-101 (acetic acid)) is the levorotatory isomer of a natural product Gossypol. AT-101 is determined to bind to Bcl-2, Mcl-1 and Bcl-xL proteins with Kis of 260±30 nM, 170±10 nM, and 480±40 nM, respectively.

(R)-(-)-Gossypol acetic acid (AT-101 (acetic acid)) is the levorotatory isomer of a natural product Gossypol. AT-101 is determined to bind to Bcl-2, Mcl-1 and Bcl-xL proteins with Kis of 260±30 nM, 170±10 nM, and 480±40 nM, respectively.

Specifications

Chemical Name or Material (R)-(-)-Gossypol acetic acid
Physical Form Solid
CAS 866541-93-7
Color Dark Brown
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C32H34O10
For Use With (Application) Cancer-programmed cell death
Synonym AT-101 (acetic acid) (-)-Gossypol acetic acid (R)-Gossypol acetic acid
Solubility Information DMSO : 25 mg/mL (43.21 mM; Need ultrasonic)
SMILES OC1=C2C(C=O)=C(O)C(O)=C(C(C)C)C2=CC(C)=[C@]1[C@]3=C(C)C=C4C(C(C)C)=C(O)C(O)=C(C=O)C4=C3O.CC(O)=O.[R]
Molecular Weight (g/mol) 578.61
Formula Weight 578.61
Percent Purity 96.98%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.