missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-Gossypol acetic acid, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
(R)-(-)-Gossypol acetic acid (AT-101 (acetic acid)) is the levorotatory isomer of a natural product Gossypol. AT-101 is determined to bind to Bcl-2, Mcl-1 and Bcl-xL proteins with Kis of 260±30 nM, 170±10 nM, and 480±40 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | (R)-(-)-Gossypol acetic acid |
| Physical Form | Solid |
| CAS | 866541-93-7 |
| Color | Dark Brown |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C32H34O10 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | AT-101 (acetic acid) (-)-Gossypol acetic acid (R)-Gossypol acetic acid |
| Solubility Information | DMSO : 25 mg/mL (43.21 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction