missing translation for 'onlineSavingsMsg'
Learn More

(R)-Flurbiprofen, TRC

Product Code. 30391807
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391807 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391807 Supplier Toronto Research Chemicals Supplier No. TRCF598730250MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-Flurbiprofen
CAS 51543-40-9
Molecular Formula C15 H13 F O2
Quantity 250 mg
Synonym (αR)-2-Fluoro-α-methyl-[1,1'-biphenyl]-4-acetic Acid, [1,1'-Biphenyl]-4-acetic acid, 2-fluoro-α-methyl-, (R)-, (-)-(R)-Flurbiprofen, (-)-Flurbiprofen, (R)-(-)-2-(2-Fluorobiphenyl-4-yl)propionic acid, (R)-Flurbiprofen, (R)-Flurbiprofen, (αR)-2-Fluoro-α-methyl[1,1'-biphenyl]-4-acetic acid, E 7869, Flurizan, MPC 7869, R-(-)-Flurbiprofen, Tarenflurbil, l-Flurbiprofen
InChI Formula InChI=1S/C15H13FO2/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11/h2-10H,1H3,(H,17,18)/t10-/m1/s1
SMILES C[C@@H](C(=O)O)c1ccc(c(F)c1)c2ccccc2
IUPAC Name (2R)-2-(3-fluoro-4-phenylphenyl)propanoic acid
Molecular Weight (g/mol) 244.26
Formula Weight 244.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.